2-[9,10-Di(naphthalen-2-yl)anthracen-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₄₀H₃₃BO₂ |
MDL Number:
MFCD26401780 CAS Number: 624744-67-8 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...