Stampa…

4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3,3',5,5'-Tetraisopropyl-[1,1'-biphenyl]-4,4'-diol, Propofol Related Compound A, 3,3',5,5'-Tetraisopropylbiphenyl-4,4'-diol

D5281-200MG D5281-1G
TCIAD5281-200MGEA 84 EUR
TCIAD5281-200MG TCIAD5281-1G
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl ≥97.0% (by GC)
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl
Formula: C₂₄H₃₄O₂
Peso molecolare: 354,53 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD20257818
CAS Number: 2416-95-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6f4bb063c86c999d7eaef8e86f0806c42f75e91&vid=d6f4bb063c86c999d7eaef8e86f0806c42f75e91&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR