Stampa…

Diethyl (2-Chlorobenzyl)phosphonate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Diethyl 2-Chlorobenzylphosphonate, (2-Chlorobenzyl)phosphonic acid Diethyl ester

D5265-1G D5265-5G
TCIAD5265-1GEA 49 EUR
TCIAD5265-1G TCIAD5265-5G
Diethyl (2-Chlorobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (2-Chlorobenzyl)phosphonate
Formula: C₁₁H₁₆ClO₃P
Peso molecolare: 262,67 g/mol
Punto di ebollizione: 146°C
MDL Number: MFCD22574983
CAS Number: 29074-98-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc1613e6fe8aa7b47702308383d7c3b7c4368ccd&vid=fc1613e6fe8aa7b47702308383d7c3b7c4368ccd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
IR to pass test
NMR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR