1,3-Dibromo-5-n-octylbenzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D5257-1G
D5257-5G
TCIAD5257-1GEA
67
EUR
TCIAD5257-1G
TCIAD5257-5G
1,3-Dibromo-5-n-octylbenzene ≥98.0% (by GC)
1,3-Dibromo-5-n-octylbenzene
Formula:
C₁₄H₂₀Br₂ Peso molecolare: 348,12 g/mol |
CAS Number:
75894-99-4 |
Risultati del test di specificazione
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...