1,1'-Biphenyl-2,2'-diamine ≥99.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,2'-Diaminobiphenyl, [1,1'-Biphenyl]-2,2'-diamine
D5255-1G
D5255-5G
TCIAD5255-1GEA
141
EUR
TCIAD5255-1G
TCIAD5255-5G
1,1'-Biphenyl-2,2'-diamine ≥99.0% (by GC, titration analysis)
1,1'-Biphenyl-2,2'-diamine
Formula:
C₁₂H₁₂N₂ Peso molecolare: 184,24 g/mol Punto di ebollizione: 151°C |
MDL Number:
MFCD00034060 CAS Number: 1454-80-4 |
Risultati del test di specificazione
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...