1,4-Diazabicyclo[2.2.2]octane dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
D5251-1G
D5251-5G
TCIAD5251-1GEA
66
EUR
TCIAD5251-1G
TCIAD5251-5G
1,4-Diazabicyclo[2.2.2]octane dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
1,4-Diazabicyclo[2.2.2]octane dihydrochloride
Formula:
C₆H₁₂N₂·₂HCl Peso molecolare: 185,09 g/mol Punto di fusione: 320°C |
CAS Number:
49563-87-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...