1,4-Diazabicyclo[2.2.2]octane dihydrobromide ≥98.0% (by total nitrogen and titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₁₂N₂·₂HBr Punto di fusione: 319°C |
MDL Number:
MFCD30725750 CAS Number: 54581-69-0 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...