Dodecahydrodibenzofuran (mixture of isomers) ≥98.0% (by GC)
Fornitore: TCI
D5232-5G
D5232-25G
TCIAD5232-5GEA
124
EUR
TCIAD5232-5G
TCIAD5232-25G
Dodecahydrodibenzofuran (mixture of isomers) ≥98.0% (by GC)
Dodecahydrodibenzofuran (mixture of isomers)
Formula:
C₁₂H₂₀O Punto di ebollizione: 259°C |
CAS Number:
13054-98-3 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...