Stampa…

Diisopropyl Phosphorochloridate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Diisopropyl Chlorophosphate, (Chlorodiisopropoxy)phosphine oxide, Phosphorochloridic acid Diisopropyl ester

D5206-1G D5206-5G
TCIAD5206-1GEA 37 EUR
TCIAD5206-1G TCIAD5206-5G
Diisopropyl Phosphorochloridate ≥98.0% (by GC, titration analysis)
Diisopropyl Phosphorochloridate
Formula: C₆H₁₄ClO₃P
Peso molecolare: 200,60 g/mol
Punto di ebollizione: 92°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00232957
CAS Number: 2574-25-6
EINECS: 624-870-1
UN: 2927
ADR: 6.1,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific gravity (20/20) 1.1030 to 1.1130

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR