5-Nitro-2,4-xylidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,4-Dimethyl-5-nitroaniline, 5-Nitro-2,4-xylidine
D5196-1G
D5196-5G
TCIAD5196-1GEA
34
EUR
TCIAD5196-1G
TCIAD5196-5G
5-Nitro-2,4-xylidine ≥98.0% (by GC, titration analysis)
5-Nitro-2,4-xylidine
Formula:
C₈H₁₀N₂O₂ Peso molecolare: 166,1800 g/mol |
MDL Number:
MFCD00025199 CAS Number: 2124-47-2 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...