Stampa…

3,5-Dichloro-4-iodopyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D5195-1G D5195-5G
TCIAD5195-1GEA 28 EUR
TCIAD5195-1G TCIAD5195-5G
3,5-Dichloro-4-iodopyridine ≥98.0% (by GC, titration analysis)
3,5-Dichloro-4-iodopyridine
Formula: C₅H₂Cl₂IN
Peso molecolare: 273,8866 g/mol
MDL Number: MFCD03094687
CAS Number: 343781-41-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fec9cf3f3ad03214e64eaa01c0e33592a7b5de6c&vid=fec9cf3f3ad03214e64eaa01c0e33592a7b5de6c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 187.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR