Stampa…

3-(Difluoromethoxy)benzylamine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (3-(Difluoromethoxy)phenyl)methanamine

D5164-1G D5164-5G
TCIAD5164-1GEA 85 EUR
TCIAD5164-1G TCIAD5164-5G
3-(Difluoromethoxy)benzylamine ≥98.0% (by GC)
3-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
Peso molecolare: 173,16 g/mol
MDL Number: MFCD00236230
CAS Number: 244022-71-7
UN: 2735
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR