Stampa…

(2,4-Dichlorobenzyl)methylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2,4-Dichloro-N-methylbenzylamine, N-Methyl-2,4-dichlorobenzylamine

D5147-1G D5147-5G
TCIAD5147-1GEA 42 EUR
TCIAD5147-1G TCIAD5147-5G
(2,4-Dichlorobenzyl)methylamine ≥98.0% (by GC, titration analysis)
(2,4-Dichlorobenzyl)methylamine
Formula: C₈H₉Cl₂N
Peso molecolare: 190,07 g/mol
Punto di ebollizione: 123°C
MDL Number: MFCD00045185
CAS Number: 5013-77-4
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c614575616d493be4014c6a88b76c626de6441b1&vid=c614575616d493be4014c6a88b76c626de6441b1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR