Stampa…

4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-propylbiphenyl, 4-(Difluoro(3,4,5-trifluorophenoxy)methyl)-3,5-difluoro-4'-propyl-1,1'-biphenyl

D5130-1G D5130-5G
TCIAD5130-1GEA 57 EUR
TCIAD5130-1G TCIAD5130-5G
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl ≥98.0% (by GC)
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl
Formula: C₂₂H₁₅F₇O
Peso molecolare: 428,35 g/mol
Punto di fusione: 49°C
MDL Number: MFCD18451990
CAS Number: 303186-20-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=379581db9cbcb220f284a3ebc5a6f206e38d70e2&vid=379581db9cbcb220f284a3ebc5a6f206e38d70e2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR