Stampa…

5,11-Dihydroindolo[3,2-b]carbazole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Indolo[3,2-b]carbazole

D5124-200MG D5124-1G
TCIAD5124-200MGEA 54 EUR
TCIAD5124-200MG TCIAD5124-1G
5,11-Dihydroindolo[3,2-b]carbazole ≥98.0% (by HPLC, total nitrogen)
5,11-Dihydroindolo[3,2-b]carbazole
Formula: C₁₈H₁₂N₂
Peso molecolare: 256,31 g/mol
Punto di fusione: 460°C
MDL Number: MFCD09879258
CAS Number: 6336-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92961135b60efdba56a18068801804fc35525583&vid=92961135b60efdba56a18068801804fc35525583&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR