N,N-Diethylbenzamide ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D5123-25G
D5123-100G
TCIAD5123-25GEA
53
EUR
TCIAD5123-25G
TCIAD5123-100G
N,N-Diethylbenzamide ≥95.0% (by GC)
N,N-Diethylbenzamide
Formula:
C₁₁H₁₅NO Peso molecolare: 177,25 g/mol Punto di ebollizione: 280°C |
MDL Number:
MFCD00026726 CAS Number: 1696-17-9 EINECS: 216-912-9 |
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 95.0 % |