1-(2,6-Dimethylphenoxy)-2-propanone ≥95.0% (by GC)
Fornitore: TCI
D5121-1G
D5121-5G
TCIAD5121-1GEA
53
EUR
TCIAD5121-1G
TCIAD5121-5G
1-(2,6-Dimethylphenoxy)-2-propanone ≥95.0% (by GC)
1-(2,6-Dimethylphenoxy)-2-propanone
Formula:
C₁₁H₁₄O₂ Punto di ebollizione: 113°C |
MDL Number:
MFCD00130266 CAS Number: 53012-41-2 EINECS: 258-292-2 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...