Stampa…

2,3-Dibromo-5-picoline ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 2,3-Dibromo-5-methylpyridine

D5100-5G D5100-25G
TCIAD5100-5GEA 68 EUR
TCIAD5100-5G TCIAD5100-25G
2,3-Dibromo-5-picoline ≥98.0% (by GC)
2,3-Dibromo-5-picoline
Formula: C₆H₅Br₂N
Peso molecolare: 250.92 g/mol
MDL Number: MFCD04112574
CAS Number: 29232-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb81cd5b4cb2cb1c7e2ac1233020d01530810cc1&vid=cb81cd5b4cb2cb1c7e2ac1233020d01530810cc1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR