2,6-Diethynylpyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D5097-200MG
D5097-1G
TCIAD5097-200MGEA
46
EUR
TCIAD5097-200MG
TCIAD5097-1G
2,6-Diethynylpyridine ≥98.0% (by GC)
2,6-Diethynylpyridine
Formula:
C₉H₅N Peso molecolare: 127,15 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD09039127 CAS Number: 75867-46-8 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...