Stampa…

Diethyl (Ethoxymethyl)phosphonate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (Ethoxymethyl)phosphonic acid Diethyl ester

D5095-5G D5095-25G
TCIAD5095-5GEA 75 EUR
TCIAD5095-5G TCIAD5095-25G
Diethyl (Ethoxymethyl)phosphonate ≥98.0% (by GC)
Diethyl (Ethoxymethyl)phosphonate
Formula: C₇H₁₇O₄P
Peso molecolare: 196,18 g/mol
Punto di ebollizione: 103°C
CAS Number: 10419-80-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR