Stampa…

1,3-Diaminopropane dihydroiodide ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3-Propanediamine dihydroiodide, Trimethylenediamine dihydroiodide

D5091-1G D5091-5G
TCIAD5091-1GEA 45 EUR
TCIAD5091-1G TCIAD5091-5G
1,3-Diaminopropane dihydroiodide ≥98.0% (by total nitrogen and titration analysis)
1,3-Diaminopropane dihydroiodide
Formula: C₃H₁₀N₂·₂HI
Peso molecolare: 329,95 g/mol
CAS Number: 120675-53-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR