Stampa…

2,4-Diamino-6-methoxypyrimidine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

D5088-1G D5088-5G
TCIAD5088-1GEA 85 EUR
TCIAD5088-1G TCIAD5088-5G
2,4-Diamino-6-methoxypyrimidine ≥98.0% (by HPLC)
2,4-Diamino-6-methoxypyrimidine
Formula: C₅H₈N₄O
Peso molecolare: 140,1400 g/mol
MDL Number: MFCD02091088
CAS Number: 3270-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58b48abb5d7768c8e9c48016fdddf88c5a9ee853&vid=58b48abb5d7768c8e9c48016fdddf88c5a9ee853&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR