2,3-Dichloro-4-pyridineboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2,3-Dichloropyridine-4-boronic acid, 2,3-Dichloro-4-pyridylboronic acid
Formula:
C₅H₄BCl₂NO₂ Punto di fusione: 141°C |
MDL Number:
MFCD04038756 CAS Number: 951677-39-7 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | gt. 97.0 and le 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...