Stampa…

2,4-Dichloro-3-formylpyridine ≥98.0% (by GC)

Fornitore: TCI
Attenzione

D5076-1G D5076-5G
TCIAD5076-1GEA 87 EUR
TCIAD5076-1G TCIAD5076-5G
2,4-Dichloro-3-formylpyridine ≥98.0% (by GC)
2,4-Dichloro-3-formylpyridine
Formula: C₆H₃Cl₂NO
MDL Number: MFCD07437909
CAS Number: 134031-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=def09b8ee40cc9014044d0352994b4bf3ab4edec&vid=def09b8ee40cc9014044d0352994b4bf3ab4edec&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR