1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2-(3,4-Dimethylphenyl)-5-methyl-1H-pyrazol-3(2H)-one
D5064-1G
D5064-5G
TCIAD5064-1GEA
64
EUR
TCIAD5064-1G
TCIAD5064-5G
1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone ≥97.0% (by GC, titration analysis)
1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone
Formula:
C₁₂H₁₄N₂O Peso molecolare: 202,26 g/mol |
MDL Number:
MFCD09795980 CAS Number: 277299-70-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...