Stampa…

2,6-Dichloro-3-iodopyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

D5061-1G D5061-5G
TCIAD5061-1GEA 50 EUR
TCIAD5061-1G TCIAD5061-5G
2,6-Dichloro-3-iodopyridine ≥98.0% (by GC)
2,6-Dichloro-3-iodopyridine
Formula: C₅H₂Cl₂IN
Peso molecolare: 273,88 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD03094686
CAS Number: 148493-37-2
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR