2,4-Dimethoxy-beta-methylcinnamic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(2,4-Dimethoxyphenyl)-2-butenoic acid
D5047-1G
D5047-5G
TCIAD5047-1GEA
54
EUR
TCIAD5047-1G
TCIAD5047-5G
2,4-Dimethoxy-beta-methylcinnamic acid ≥98.0% (by GC, titration analysis)
2,4-Dimethoxy-beta-methylcinnamic acid
Formula:
C₁₂H₁₄O₄ Peso molecolare: 222,24 g/mol |
MDL Number:
MFCD00512029 CAS Number: 7706-67-4 Merck Index: 14,03203 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...