2-(Dicyclohexylphosphino)-2'-methylbiphenyl ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Dicyclohexyl(2'-methyl-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2'-methyl-)-1,1'-biphenyl, MePhos, 2-Dicyclohexylphosphino-2'-methylbiphenyl
D5035-1G
D5035-5G
TCIAD5035-1GEA
96
EUR
TCIAD5035-1G
TCIAD5035-5G
2-(Dicyclohexylphosphino)-2'-methylbiphenyl ≥97.0% (by GC)
2-(Dicyclohexylphosphino)-2'-methylbiphenyl
Formula:
C₂₅H₃₃P Peso molecolare: 364,513 g/mol |
MDL Number:
MFCD03094577 CAS Number: 251320-86-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...