Stampa…

2,7-Diiodo-9,9-dimethylfluorene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,7-Diiodo-9,9-dimethyl-9H-fluorene

D5032-1G D5032-5G
TCIAD5032-1GEA 62 EUR
TCIAD5032-1G TCIAD5032-5G
2,7-Diiodo-9,9-dimethylfluorene ≥98.0% (by GC)
2,7-Diiodo-9,9-dimethylfluorene
Formula: C₁₅H₁₂I₂
Peso molecolare: 446,07 g/mol
MDL Number: MFCD12827985
CAS Number: 144981-86-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR