Stampa…

3,5-Dibromo-1H-1,2,4-triazole ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,5-Dibromo-1,2,4-triazole

D5030-5G D5030-25G
TCIAD5030-5GEA 63 EUR
TCIAD5030-5G TCIAD5030-25G
3,5-Dibromo-1H-1,2,4-triazole ≥97.0% (by GC)
3,5-Dibromo-1H-1,2,4-triazole
Formula: C₂HBr₂N₃
Peso molecolare: 226,8600 g/mol
MDL Number: MFCD00487260
CAS Number: 7411-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61a9c48c968efd357e982d2b408d1a6771e1bd26&vid=61a9c48c968efd357e982d2b408d1a6771e1bd26&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR