4,4''-Dibromo-1,1':3',1''-terphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4,4''-Dibromo-m-terphenyl
D5023-200MG
D5023-1G
TCIAD5023-200MGEA
59
EUR
TCIAD5023-200MG
TCIAD5023-1G
4,4''-Dibromo-1,1':3',1''-terphenyl ≥98.0% (by GC)
4,4''-Dibromo-1,1':3',1''-terphenyl
Formula:
C₁₈H₁₂Br₂ Peso molecolare: 388,10 g/mol |
MDL Number:
MFCD28138091 CAS Number: 83909-22-2 UN: 3152 ADR: 9,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...