Stampa…

3,5-Difluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,5-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Difluorophenylboronic acid Pinacol ester

D5017-1G D5017-5G
TCIAD5017-1GEA 37 EUR
TCIAD5017-1G TCIAD5017-5G
3,5-Difluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,5-Difluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BF₂O₂
Peso molecolare: 240,06 g/mol
MDL Number: MFCD11504963
CAS Number: 863868-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2960dc622b691cf2f844ca32d5da8e899b3580e9&vid=2960dc622b691cf2f844ca32d5da8e899b3580e9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR