7-Diethylamino-3-phenylcoumarin ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₉H₁₉NO₂ |
MDL Number:
MFCD00408422 CAS Number: 84865-19-0 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...