Stampa…

3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,6-Dibromo-9-(4-(tert-butyl)phenyl)-9H-carbazole

D5000-200MG D5000-1G
TCIAD5000-200MGEA 36 EUR
TCIAD5000-200MG TCIAD5000-1G
3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole
Formula: C₂₂H₁₉Br₂N
Peso molecolare: 457,21 g/mol
MDL Number: MFCD12024283
CAS Number: 741293-42-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR