Stampa…

4,4''-Dibromo-1,1':2',1''-terphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4,4''-Dibromo-o-terphenyl

D4998-200MG D4998-1G
TCIAD4998-200MGEA 79 EUR
TCIAD4998-200MG TCIAD4998-1G
4,4''-Dibromo-1,1':2',1''-terphenyl ≥98.0% (by GC)
4,4''-Dibromo-1,1':2',1''-terphenyl
Formula: C₁₈H₁₂Br₂
Peso molecolare: 388,10 g/mol
MDL Number: MFCD28138092
CAS Number: 24253-43-8
UN: 3152
ADR: 9,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 170.0 to 174.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR