Stampa…

3,3''-Dibromo-1,1':2',1''-terphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3,3''-Dibromo-o-terphenyl

D4997-200MG D4997-1G
TCIAD4997-200MGEA 98 EUR
TCIAD4997-200MG TCIAD4997-1G
3,3''-Dibromo-1,1':2',1''-terphenyl ≥98.0% (by GC)
3,3''-Dibromo-1,1':2',1''-terphenyl
Formula: C₁₈H₁₂Br₂
Peso molecolare: 388,10 g/mol
MDL Number: MFCD28138090
CAS Number: 95918-90-4
UN: 3152
ADR: 9,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR