Stampa…

(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (S)-MONOPHOS

D4993-1G D4993-5G
TCIAD4993-1GEA 57 EUR
TCIAD4993-1G TCIAD4993-5G
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98.0% (by HPLC, titration analysis)
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine
Formula: C₂₂H₁₈NO₂P
Peso molecolare: 359,3576 g/mol
MDL Number: MFCD03426988
CAS Number: 185449-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fcb61642d81ec61f1210f68e5524e39b9483f2&vid=69fcb61642d81ec61f1210f68e5524e39b9483f2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +560 to +600 deg (C=0.3,CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR