Stampa…

9-(9,9-Dimethylfluoren-2-yl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

D4981-1G D4981-5G
TCIAD4981-1GEA 156 EUR
TCIAD4981-1G TCIAD4981-5G
9-(9,9-Dimethylfluoren-2-yl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
9-(9,9-Dimethylfluoren-2-yl)-9H-carbazole
Formula: C₂₇H₂₁N
Peso molecolare: 359,47 g/mol
CAS Number: 866100-16-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR