Stampa…

3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D4979-200MG D4979-1G
TCIAD4979-200MGEA 67 EUR
TCIAD4979-200MG TCIAD4979-1G
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole
Formula: C₂₄H₁₅Br₂N
Peso molecolare: 477,20 g/mol
MDL Number: MFCD22571691
CAS Number: 1345021-52-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR