Stampa…

9,10-Dibromo-2-methylanthracene ≥96.0% (by GC)

Fornitore: TCI EUROPE

D4977-200MG D4977-1G
TCIAD4977-200MGEA 78 EUR
TCIAD4977-200MG TCIAD4977-1G
9,10-Dibromo-2-methylanthracene ≥96.0% (by GC)
9,10-Dibromo-2-methylanthracene
Formula: C₁₅H₁₀Br₂
Peso molecolare: 350,05 g/mol
MDL Number: MFCD00266492
CAS Number: 177839-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=302e4f5eb50482458639c3793ece77b0ff3067fb&vid=302e4f5eb50482458639c3793ece77b0ff3067fb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR