Stampa…

(Dimethanesulphinylidene)amine ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Dimethylsulphoximine, (S-Methylsulphonimidoyl)methane

D4971-1G D4971-5G
TCIAD4971-1GEA 50 EUR
TCIAD4971-1G TCIAD4971-5G
(Dimethanesulphinylidene)amine ≥98.0% (by GC)
(Dimethanesulphinylidene)amine

Dimethylsulfoximine [D4971] is a building block for the synthesis of sulfoximines and utilized in various types of sulfoximidation reactions.


Possible applications include such as SN2 reactions, nucleophilic attacks to carbonyl groups and cross-coupling reactions, and providing the target compounds in high yields under various conditions. Furthermore, this functional group can be inserted at a late stage of complex syntheses via C-H activation.

Formula: C₂H₇NOS
Peso molecolare: 93,15 g/mol
Punto di ebollizione: 100°C
MDL Number: MFCD02656633
CAS Number: 1520-31-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR