Stampa…

2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-D-cytidine, (2R,3R,4R,5R)-2-(4-Amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-5-methyltetrahydrofuran-3,4-diyl diacetate

D4969-1G D4969-5G D4969-25G
TCIAD4969-1GEA 35 EUR
TCIAD4969-1G TCIAD4969-5G TCIAD4969-25G
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine
Formula: C₁₃H₁₆FN₃O₆
Peso molecolare: 329,28 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD07369302
CAS Number: 161599-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5ba82de767c096485a4dbc22a46ca33401c40b1&vid=c5ba82de767c096485a4dbc22a46ca33401c40b1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 187.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR