6,8-Dibromoimidazo[1,2-a]pyrazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D4967-1G
D4967-5G
TCIAD4967-1GEA
37
EUR
TCIAD4967-1G
TCIAD4967-5G
6,8-Dibromoimidazo[1,2-a]pyrazine ≥98.0% (by GC)
6,8-Dibromoimidazo[1,2-a]pyrazine
Formula:
C₆H₃Br₂N₃ Peso molecolare: 276,92 g/mol |
MDL Number:
MFCD08460056 CAS Number: 63744-22-9 |
Risultati del test di specificazione
Appearance | White to Amber powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...