Stampa…

1-(2,6-Dimethylphenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-(2,6-Dimethylphenyl)thiourea, (2,6-Dimethylphenyl)thiourea, 1-(2,6-Xylyl)thiourea, N-(2,6-Dimethylphenyl)thiourea, 2,6-Dimethylphenylthiourea

D4958-5G D4958-25G
TCIAD4958-5GEA 101 EUR
TCIAD4958-5G TCIAD4958-25G
1-(2,6-Dimethylphenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-(2,6-Dimethylphenyl)-2-thiourea
Formula: (C₆H₃(CH₃)₂)NC(S)NH₂
Peso molecolare: 180,27 g/mol
MDL Number: MFCD00041165
CAS Number: 6396-76-5
EINECS: 229-005-8
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR