α-Bromo-4-(difluoromethoxy)toluene ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
1-(Bromomethyl)-4-(difluoromethoxy)benzene, p-(Difluoromethoxy)benzyl bromide, 4-(Difluoromethoxy)benzyl bromide
Formula:
C₈H₇BrF₂O Peso molecolare: 237.04 g/mol Punto di ebollizione: 52°C |
MDL Number:
MFCD00085007 CAS Number: 3447-53-8 UN: 3265 ADR: 8 II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...