Stampa…

4-(Difluoromethoxy)benzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

D4955-1G D4955-5G
TCIAD4955-1GEA 33 EUR
TCIAD4955-1G TCIAD4955-5G
4-(Difluoromethoxy)benzonitrile ≥98.0% (by GC)
4-(Difluoromethoxy)benzonitrile
Formula: C₈H₅F₂NO
Peso molecolare: 169,1282 g/mol
MDL Number: MFCD00085006
CAS Number: 90446-25-6
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR