3,5-Di(1-pyrenyl)pyridine ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
D4931-200MG
D4931-1G
TCIAD4931-200MGEA
69
EUR
TCIAD4931-200MG
TCIAD4931-1G
3,5-Di(1-pyrenyl)pyridine ≥98.0% (by HPLC, total nitrogen)
3,5-Di(1-pyrenyl)pyridine
Formula:
C₃₇H₂₁N Peso molecolare: 479,58 g/mol |
CAS Number:
1246467-58-2 |
Risultati del test di specificazione
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...