N,N'-Diphenyl-N,N'-di(m-tolyl)-1,4-phenylenediamine ≥98.0% (by GC)
Fornitore: TCI EUROPE
D4930-1G
D4930-5G
TCIAD4930-1GEA
81
EUR
TCIAD4930-1G
TCIAD4930-5G
N,N'-Diphenyl-N,N'-di(m-tolyl)-1,4-phenylenediamine ≥98.0% (by GC)
N,N'-Diphenyl-N,N'-di(m-tolyl)-1,4-phenylenediamine
Formula:
C₃₂H₂₈N₂ Peso molecolare: 440,59 g/mol |
CAS Number:
80223-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...