Stampa…

N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

D4929-1G D4929-5G
TCIAD4929-1GEA 61 EUR
TCIAD4929-1G TCIAD4929-5G
N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine ≥98.0% (by HPLC, total nitrogen)
N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine
Formula: C₃₈H₂₈N₂
Peso molecolare: 512,66 g/mol
MDL Number: MFCD28291914
CAS Number: 139994-47-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 192.0 to 196.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR