Stampa…

5,6-Dichloro-1H-indole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5,6-Dichloroindole

D4924-1G D4924-5G
TCIAD4924-1GEA 189 EUR
TCIAD4924-1G TCIAD4924-5G
5,6-Dichloro-1H-indole ≥98.0% (by GC)
5,6-Dichloro-1H-indole
Formula: C₈H₅Cl₂N
Peso molecolare: 186,0400 g/mol
MDL Number: MFCD01075036
CAS Number: 121859-57-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR